Mag Chips
Supplier: NADA SCIENTIFIC LTD.
B2637-115
470110-256EA 100.5 CAD
470110-256
Mag Chips
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Easier to Use and Clean Up than Iron Filings.
  • They are 2mm-long pieces of zinc-plated iron wire
  • When exposed to a magnetic field, they align, exhibiting the field lines clearly. 50g container