Sterile Spec-Tec™ Stretch Glove Liners, Wells Lamont
Supplier: Wells Lamont
M104L M104M M104S M104XL
80091-128PK 1228.06 CAD
80091-128 80091-130 80091-132 80091-134
Sterile Spec-Tec™ Stretch Glove Liners, Wells Lamont
Gloves
These glove liners feature high performance fiber blended with Lycra® for stretch and reusability.
  • The gloves feature good tactile sensitivity
Ambidextrous and latex free, the gloves can be washed 12 times without shrinking.
Certifications: ANSI Level 2 Cut Protection.