SP Bel-Art Portable Vacuum Aspirator Collection Systems, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19917-0150 F19917-0250
89171-442EA 1598.57 CAD
89171-442 10025-014
SP Bel-Art Portable Vacuum Aspirator Collection Systems, Bel-Art Products, a part of SP
Pumps Vacuum Aspiration Systems
This compact, portable system is ideal for vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes and is available with the original strength or HiFlow pump.
  • Lightweight
  • Compact and easy to use
  • Built-in, maintenance free pump quickly and conveniently aspirates fluids
  • Low noise level (76dbs or less at 1 meter); minimal pump vibration
  • Chemical- and corrosion-resistant
  • Heavy-walled polypropylene construction
  • Replaceable hydrophobic filter protects from fluid contamination
  • Collection bottle, lid, tubing, and connectors are all autoclavable
HiFlow model has a stronger flow rate to work reliably with up to 8-channel pipette tips, even with 4 channels empty
HiFlow model is also CE marked and CSA certified and the transformer includes an additional cord for 220V type C outlets.
HiFlow model includes quick disconnects to allow easy removal, emptying and replacement of the bottle.
Certifications: 110/120V UL listed.
Ordering information: Recommended accessories are replaceable hydrophobic filters (89171-444 and 10025-016).