Detergent-OUT™ Tween® For Removal of Polysorbate Detergents, G-Biosciences
Supplier: G-Biosciences
786-214 786-215
CA71002-884EA 736.93 CAD
CA71002-884 CA71002-886
Detergent-OUT™ Tween® For Removal of Polysorbate Detergents, G-Biosciences
Cleaning Agents Decontaminants
G-Biosciences offers three DetergentOUT™ detergent removal systems for the removal of detergents from peptide and protein solutions
DetergentOUT™ Tween® removes polysorbate detergents without significant loss of proteins, dilution of the protein solution, or change to the buffer composition of the protein solution.
The binding capacity is approximately 85mg polysorbate detergent per 1mL resin. The resin does not bind to protein, allowing for >95% protein recovery. DetergentOUT™ Tween® resin binds other commonly used detergents, but with lower affinity compared to G-Biosciences' DetergentOUT™ GBS10.
The DetergentOUT™ Tween® is a spin column format detergent removal resin that offers higher binding affinity for polysorbate detergents, commercially known as Tween®
Ordering information: Accessories available.