RIPA Lysis and Extraction Buffer, G-Biosciences
Supplier: G-Biosciences
G-Biosciences' RIPA Lysis and Extraction Buffer is a complete lysis buffer for the release of cytoplasmic, membrane and nuclear proteins from adherent and suspension cultured mammalian cells
RIPA Lysis Buffer does not contain protease inhibitors; however, it is fully compatible with G-Biosciences' range of individual protease inhibitors and cocktails.
The RIPA lysis buffer is fully compatible with many applications, including reporter assays, protein assays, immunoassays and other protein purification techniques
Ordering information: Accessories available.