SP Bel-Art 80-Well Reversible PCR and Microcentrifuge Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18845-0001 F18845-0012
89230-648CS 558.27 CAD
89230-648 89230-652 89230-654 89230-660 89230-662 89230-670 89230-672 89230-674 89230-676
SP Bel-Art 80-Well Reversible PCR and Microcentrifuge Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Perfect for benchtop use or freezer storage, this double-sided polypropylene plastic rack accommodates 0.2 ml thin-walled PCR tubes on one side while the opposite side holds 1.5 ml to 2.0 ml microcentrifuge tubes.
  • Ideal compact storage solution
  • Molded-in handles
  • Color-coded racks assist in quick visual identification
  • Can be used for freezer storage
  • Autoclavable
80 wells on each side keep tubes upright in a 5 x 16 array and are numbered for easy sample identification. Twelve rack colors assist in sample organization and quick visual identification.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable