DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Open Wrists and Ankles, Bound Seams
Supplier: DuPont
QC120BYLMD001200 QC120BYLLG001200 QC120BYLXL001200 QC120BYL3X001200 QC120BYL4X001200 QC120BYL2X001200 QC120BYL5X001200 QC120BYL6X001200
CA10010-224CS 1158.71 CAD
CA10010-224 CA10010-226 CA10010-228 CA10010-232 CA10010-234 10016-360 10016-362 10016-364
DuPont™ Tychem® 2000 Coveralls with Laydown Collar and Open Wrists and Ankles, Bound Seams
Disposable Garments
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
  • Laydown collar
  • Open wrists and ankles
  • Storm flap with adhesive closure
  • Bound seams
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Tightly sewn seams areMore Product Information
Order Now
SPECIFICATIONS
Garment application Cleanroom,Laboratory