Warehouse Signs without Header, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
M372AB M372RB M436PB M436PC M436RB M436RC
94017-086EA 31.85 CAD
94017-086 94017-088 94017-116 94017-118 94017-146 94017-148 94017-150 94017-152 94017-218 94017-220 94017-222 94017-224
Warehouse Signs without Header, National Marker
Signs
Safety signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
These signs point out easily overlooked dangers in and around the workplace. Avoid accidents and keep employees safe by clearly identifying hazardous equipment situations. This can include heavy equipment, moving machine parts and powered vehicles.