Miniature Circuit Breaker Lockout - Pin Out Standard, Brady Worldwide®
Supplier: Brady
90848 90844 90847 90848 90850 90851 90845 90845 90850 90844 90847 90851
CA89007-890EA 19.53 CAD
89007-882 CA89007-890 89007-892 89007-890 89007-886 CA89007-884 CA89007-882 89007-884 CA89007-892 CA89007-888 89007-888 CA89007-886
Miniature Circuit Breaker Lockout - Pin Out Standard, Brady Worldwide®
Security Locks
Miniature Circuit Breaker Lockout - Pin Out Standard For use on miniature circuit breakers
Miniature Breaker Lockouts require NO TOOLS to install. Standard "Pin" style lockouts install easily with the aid of a push button. Lockouts are available for single and multi-pole breakers. Recommended to lock with Brady Safety padlocks (Ex: 89008-058 - Red) or other lightweight padlock with shackle diameter up to 9/32". Lockouts are made of glass-filled nylon.