Sodium octanoate ≥90%

Supplier: Spectrum Chemical Mfg. Corp.
Warning

SO145-12KG SO145-45KGBL
CA700005-320EA 9476 CAD
CA700005-320 CA700005-324
Sodium octanoate ≥90%
Sodium octanoate

Sodium Caprylate is the sodium salt C₈H₁₅O₂Na of caprylic acid used especially in the topical treatment of fungal infections.


In cosmetics and personal care, it is applied as a surfactant and cleansing agent.

Formula: C₈H₁₅NaO₂
Melting Pt: 225…355 °C
Density: >0.9949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR