First Aid and Spill Response Signs, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
EGA1PB EGA1R EGA1RB EGA2AB EGA2P EGA2PB EGA2R EGA2RB EGA3AB EGA3P EGA3PB EGA3R EGA3RB EGA4AB EGA4P EGA4PB EGA4R EGA4RB SF116AB SF116P SF116PB SF116R SF116RB SF160AB SF160PB SF160RB SF161AB SF161PB SF161RB SF175AB SF175PB SF175RB SF176AB SF176PB SF176RB SF41P SF41PB SF41R SF41RB SF43PB SF43RB SF45PB SF45R SF45RB SF47P SF47R SF47RB SFA1P SFA1R SFA3P SFA3R SFA5P SFA5R SGA1AP
94012-792EA 48.03 CAD
94012-792 94012-794 94012-796 94012-798 94012-800 94012-802 94012-804 94012-806 94012-808 94012-810 94012-812 94012-814 94012-816 94012-818 94012-820 94012-822 94012-824 94012-826 94018-106 94018-108 94018-110 94020-898 94020-900 94020-902 94020-904 94020-906 94021-010 94021-012 94021-014 94021-016 94021-018 94021-020 94021-022 94021-024 94021-026 94021-028 94021-030 94021-032 94021-106 94021-108 94021-110 94021-112 94021-114 94021-116 94021-136 94021-138 94021-140 94021-142 94021-184 94021-186 94021-188 94021-190 94021-192 94021-194 94021-196 94021-198 94021-200 94021-202 94021-204 94021-206 94021-250 94021-252 94021-254 94021-256 94021-258 94021-260 94021-262 94021-264 94021-266 94021-268 94021-272 94021-458 94021-460 94021-462
First Aid and Spill Response Signs, National Marker
Signs
Alert your employees to the location of first aid and emergency response stations.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes