Laboratory Bottle, Square, GL Thread, Ace Glass
Supplier: Ace Glass
5560-10 5560-15 5560-20 5560-25
89051-892CS 604.96 CAD
89051-892 89051-894 89051-896 89051-898
Laboratory Bottle, Square, GL Thread, Ace Glass
Bottles Glass Bottles
These chemically resistant square bottles are fabricated from borosilicate glass.
They are graduated, and are provided with polypropylene, blue cap, and removable pouring ring that allows drip-free operations. For use up to 140 °C. Not suitable for vacuum or pressure use. Can be hot air sterilized. Height measurement listed is with cap.