Cap, Premium, Ace Glass Incorporated
Supplier: Ace Glass
7627-02 7627-03
89199-106CS 467.12 CAD
89199-106 89199-108
Cap, Premium, Ace Glass Incorporated
Closures Bottle Closures
This premium cap and pour ring are designed for GL45 thread Duran bottles
Made of colorless, TPCh260 resin so that there are no leachables. Cap has high chemical resistance, and is leak-tight due to a PTFE coated silicone seal. Meets USP standards. Autoclavable.