Axygen® Automation Tips for Hamilton® Microlab® Star Line and Nimbus®, Corning
Supplier: Corning
HT-10-CBK-HTR HT-10-CBK-HTR-S HT-300-CBK-HTR HT-300-CBK-HTR-S HT-300CBKSTK HT-50-CBK-HTR HT-50-CBK-HTR-S HT-50-CBKSTK HTF-1000-CBK-HTR-S HTF-10-CBK-HTR-S HTF-300-CBK-HTR-S HTF-50-CBK-HTR-S HTF-10-CBK-HTR-S-5 HT-50-CBK-HTR-5 HT-50-CBK-HTR-S-5 HTF-50-CBK-HTR-S-5 HT-300-CBK-HTR-5 HTF-300-CBK-HTRS5 HT-1000-CBK-HTR-5 HT-1000-CBK-HTRS5 HTF-1000-CBKHTRS5 HT-10-CBK-HTR-5 HT-10-CBK-HTR-S-5 HT-10-C-STK HT-10-C-STK-S HT-50-C-STK HT-50-C-STK-S HT-300-C-STK HT-10-CBK-STK-S HT-50-CBK-STK-S HT-300-CBK-STK-S HT-10-C-HTR-5 HT-10-C-HTR-S-5 HTF-10-C-HTR-S-5 HT-50-C-HTR-5 HTF-50-C-HTR-S-5 HT-300-C-HTR-5 HT-300-C-HTR-S-5 HTF-300-C-HTR-S-5 HT-1000-C-HTR-5 HT-1000-C-HTR-S-5
89230-130CS 639.46 CAD
89230-130 89230-132 89230-140 89230-142 89230-148 89230-150 89230-152 89230-158 89230-160 89230-162 89230-166 89230-170 10057-858 10057-860 10057-862 10057-864 10057-866 10057-868 10057-870 10057-872 10057-874 10057-876 10060-160 10060-162 CA10159-072 CA10159-074 CA10159-076 CA10159-078 CA10159-080 CA10746-458 CA10746-460 CA10746-462 CA10127-403 CA10127-405 CA10127-407 CA10127-409 CA10127-411 CA10127-413 CA10127-415 CA10127-417 CA10127-505 CA10127-507 CA10127-509 CA10127-511
Axygen® Automation Tips for Hamilton® Microlab® Star Line and Nimbus®, Corning
Pipette Tips
Axygen brand automation tips are compatible with Hamilton® Microlab® Star and Nimbus® robotic systems.
  • Conductive black or clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
Tips are available in 4 sizes (10, 50, 300, and 1000 µl) and are packaged in either traditional hanging tip racks ('frame' style) or deck-seating nested tip racks. As with all Axygen robotic tips, they are manufactured to stringent specifications under strict process control and assembled via automation to ensure consistent performance and quality.