Bottle, Wide Mouth, Ace Glass
Supplier: Ace Glass
5559-05 5559-07 5559-09 5559-20 5559-24 5559-205 5559-207 5559-209 5559-220 5559-300 5559-304 5559-306 5559-308 5559-03
89199-062CS 766.37 CAD
89199-062 89199-064 89199-066 89199-068 89199-070 89199-072 89199-074 89199-076 89199-078 89199-080 89199-082 89199-084 89199-086 89199-088 89199-090 10767-216 10767-900
Bottle, Wide Mouth, Ace Glass
Bottles Glass Bottles
These wide-neck, graduated laboratory bottles are chemically resistant and stable.
Supplied with GLS80 quick-release polypropylene blue closure. Available clear, amber, or safety coated. Amber and clear versions are rated for use up to 140 °C; safety coated version rated for use up to 135 °C. Not suitable for vacuum or pressure use. Can be hot air sterilized or autoclaved. Fabricated from borosilicate glass.