WHEATON® Aluminum Seals for Serum Bottles, Ace Glass Incorporated
Supplier: Ace Glass
5532-27 5532-07 5532-37 5532-38 5532-39
89051-758CS 276 CAD
89051-758 89051-760 10098-762 10098-764 76106-984
WHEATON® Aluminum Seals for Serum Bottles, Ace Glass Incorporated
Closures Chromatography Vial Closures
These seals are for use with flange and slotted style stoppers.
  • Three styles: center disc tear out, complete tear off, and open top
  • For use with serum bottles and vials
  • Made of aluminum