Adapter, Distillation, Ace Glass Incorporated
Supplier: Ace Glass
5299-12 5299-07 5299-10 5299-01 5299-16 5299-03
89214-890EA 1392.12 CAD
89214-890 89214-880 89214-884 89214-882 89214-888 89214-886
Adapter, Distillation, Ace Glass Incorporated
Distillation Components Distillation Adapters
This distillation adapter is for use with bench or pilot plant reactors.
  • Center vessel collects moisture
  • With bottom PTFE plug stopcock
  • Fabricated from borosilicate glass
Moisture is collected in the center vessel and drained off through the bottom stopcock which is ground to accept a compression style fitting. Stopcock is a PTFE plug. Available with either one or two reinforced top standard taper outer joints, and single standard taper inner joint at bottom next to stopcock. Joint Size value represents the compression fitting joint size .