Economy Wash Bottle, Ace Glass
Supplier: Ace Glass
12461-07 12461-09 12461-11 12461-13
71000-184CS 738.91 CAD
71000-184 71000-186 71000-194 71000-196
Economy Wash Bottle, Ace Glass
Bottles Wash Bottles
These wash bottles come with a top-angled spout at a money-saving price.
Bottle, tubing, and tip are made of polyethylene with a polyallomer cap and nozzle. Molded tip dispenses a uniform stream. Can be cut back to increase flow.