Safety Wash Bottle, Ace Glass
Supplier: Ace Glass
12464-02 12464-06 12464-10 12464-14 12464-16 12464-18
71000-198CS 275.21 CAD
71000-198 71000-200 71000-202 71000-204 71000-206 71000-208
Safety Wash Bottle, Ace Glass
Bottles Wash Bottles
These wide mouth safety wash bottles are guaranteed to be leak-proof.
They are made of flexible, impact-resistant polyethylene with color-coded polyallomer closure and spout. Standard 'diamond alert' symbols indicate what type hazard the contents may present. Available in 500 ml (16 oz.) size only.