Reservoir Bottles, Graduated Glass, Reagent, Ace Glass Incorporated
Supplier: Ace Glass
5414-07 5414-10 5414-137 5414-139 5414-141 5414-15
89051-656EA 742.83 CAD
89051-656 89051-658 89051-662 89051-664 89051-666 89051-670
Reservoir Bottles, Graduated Glass, Reagent, Ace Glass Incorporated
Bottles Solution Bottles
This heavy-wall borosilicate glass bottle has three PTFE valves with Tefzel keys.
Pressure-tight PTFE coated Fluoron forms a seal within the cap; hence, fluid contact is restricted to glass, PTFE and Tefzel. The three individually controlled valves permit the application of gas under pressure to the bottle for venting, flushing, or delivery of the bottle contents to one or two points. Bottles may be pressurized in isolation or in series with the other bottles. One of the valves may be used to allow corrosive fumes to be vented safely. Rated to 14 psi at ambient; must be adequatelyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable