Adapter, Vacuum, Long Stem, Ace Glass Incorporated
Supplier: Ace Glass
9121-04 9121-06 9121-08
89064-996EA 168.74 CAD
89064-996 89082-822 89082-826
Adapter, Vacuum, Long Stem, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This apparatus is useful as a vacuum adapter or addition tube.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
  • With hose connection
  • Fabricated from borosilicate glass
The stem length below the joint on this adapter is 130mm. Use with a 5/16-inch I.D. tubing, size A hose connection. Adapter has an outer standard taper joint at the top and an inner standard taper joint at the bottom.