Adapter, PTFE Pour Spout, Ace Glass Incorporated
Supplier: Ace Glass
7645-04 7645-07 7645-15
89084-660EA 198.55 CAD
89084-660 89084-662 89084-664
Adapter, PTFE Pour Spout, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This PTFE threaded pour spout adapter with included FETFE o-ring prevents material contact with threads.
  • Pour spout
  • O-ring included
Adapter 89084-660 and 89084-662 may be used in conjunction with FETEF O-Ring 89060-984 and 89061-016. The FETEF O-Ring 89061-030 is in conjuction with the item 89084-664.