Gas Adapter, Ace Glass Incorporated
Supplier: Ace Glass
7809-03 7809-07
89060-466EA 274.55 CAD
89060-466 89060-468
Gas Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter features a T-Bore, 2mm stopcock at the top.
  • Bottom [ST] inner joint
  • Top T-Bore stopcock
  • Fabricated from borosilicate glass
Adapter has a standard taper inner joint at the bottom and a T-Bore, 2mm stopcock at the top. "[ST] Joint" value represents the value of the inner [ST] joint.