Boston Bottle, Round, Safety Coated, Ace Glass
Supplier: Ace Glass
5546-08 5546-10 5546-12
89051-846PK 108.62 CAD
89051-846 89051-848 89051-850 89051-852
Boston Bottle, Round, Safety Coated, Ace Glass
Bottles Glass Bottles
Clear glass bottles, fabricated of soda lime, flint glass.
  • Not autoclavable
  • Supplied without caps
PVC safety coated to reduce potential breakage, and to prevent exposure to laboratory personnel in handling hazardous materials.
Order Now
SPECIFICATIONS
Environmentally Preferable