Bottle, Wide Mouth, Ace Glass
Supplier: Ace Glass
5549-08 5549-16
89051-862PK 174.82 CAD
89051-862 89051-864
Bottle, Wide Mouth, Ace Glass
Bottles Glass Bottles
These clear glass bottles are fabricated of soda lime, flint glass.
  • Not autoclavable
  • Supplied without caps
They are PVC safety coated to reduce potential breakage, and exposure to laboratory personnel in handling hazardous materials.