Adapter, Bleed Capillary, Ace Glass Incorporated
Supplier: Ace Glass
9328-02 9328-04 9328-18 9328-22
89074-394EA 133.38 CAD
89074-394 89074-396 89074-398 89074-400
Adapter, Bleed Capillary, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This bleed capillary adapter is fabricated from borosilicate glass.
  • Top [ST] joint
  • With hose connection
  • Fabricated from borosilicate glass
The length of this adapter is measured from the top of the joint to the tip. Used with a 5/16-inch I.D. tubing, size A hose connection.
Ordering information: Contact your Avantor representative for a listing of which flasks and heads fit this adapter.