Housekeeping and Hygiene Safety First Signs, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
SF130P SF130PB SF130R SF130RB SF131AB SF131P SF131PB SF131R SF131RB SF132P SF132PB SF132R SGA2AP
94020-912EA 31.85 CAD
94020-912 94020-914 94020-916 94020-918 94020-920 94020-922 94020-924 94020-926 94020-928 94020-930 94020-932 94020-934 94020-936 94020-938 94020-940 94021-064 94021-066 94021-068 94021-274
Housekeeping and Hygiene Safety First Signs, National Marker
Signs
Help keep your facility clean and safe
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Post signs encouraging employees to keep their work areas clean and organized.