Adapter, Firestone, Ace Glass Incorporated
Supplier: Ace Glass
5258-06 5258-12 5258-16 5258-62
89051-468EA 205.44 CAD
89051-468 89051-470 89051-472 89051-474
Adapter, Firestone, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
When inserted into a flask, this rotary evaporator “splash guard” adapter combines with flask neck to give the best protection against a splash-up.
  • Top and bottom [ST] medium length joints
  • Provides best splash protection
  • Fabricated from borosilicate glass
The adapter's standard taper medium length joints, top and bottom, will accept full length standard taper joints. Overall height is kept to a minimum to effect best distillation.
Order Now
SPECIFICATIONS
Environmentally Preferable