Adapter, Firestone, with Fritted Disc, Ace Glass Incorporated
Supplier: Ace Glass
5257-43 5257-49 5257-53 5257-62
89051-460EA 272.77 CAD
89051-460 89051-462 89051-464 89051-466
Adapter, Firestone, with Fritted Disc, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This rotary evaporator “splash guard” adapter features a coarse Porosity A fritted disc that assures there will be no carry-over in the event of splash-up.
  • Top and bottom [ST] medium length joints
  • Coarse Porosity A fritted disc
  • Fabricated from borosilicate glass
The adapter's standard taper medium length joints, top and bottom, will accept full length standard taper joints. The coarse Porosity A (147-174 micron) fritted disc assures there will be no carry-over in the event of splash-up. Overall length is kept to a minimum to effect best distillation.