Fluorescent Nerve Terminal Probes and Staining Kits, Biotium
Supplier: Biotium
70019 70020 70021 70022 70023 70024 70025 70026 70027 70028 70029 70030 70031 70031-1 70032 70034 70036 70037 70038 70039 70040 70041 70042 70043 70044 70045 70050 70051
89139-396EA 781.61 CAD
89139-396 89139-398 89139-400 89139-402 89139-404 89139-406 89139-408 89139-410 89139-412 89139-414 89139-416 89139-418 89139-420 89139-422 89139-424 89139-426 89139-430 89139-432 89139-434 89139-436 89139-438 89139-440 89139-442 89139-444 89139-446 89139-448 89139-458 89139-460
Fluorescent Nerve Terminal Probes and Staining Kits, Biotium
Assays Cellular Assays
Nerve terminal probes are a series of fluorescent cationic styryl dyes developed to follow synaptic activity
Styryl dyes stain synaptic vesicles in an activity-dependent fashion. The dyes are virtually non-fluorescent in aqueous solution, but become brightly fluorescent upon insertion into the plasma membrane. During endocytosis following neuronal stimulation, the dyes become trapped in vesicles. After free dye is removed from the cell surface by washing, the remaining fluorescence is proportional to the number of newly formed endocytic vesicles. During exocytosis, the dyes are released to the aqueous medium, causing a decrease in fluorescent signal. As a result, the change in fluorescence intensity reflects the amount of endocytosis and exocytosis occuring during synaptic activity. The rate of fluorescence increase during endocytosis (on-rate) and the rate of fluorescence decrease during exocytosis (off-rate) varies from dye to dye. In general, dyes with longer lipophilic tails and more double bonds have a faster on-rate and slower off-rate. AM dyes have an additional amine group and therefore tend to be more water soluble than their FM dye counterparts, resulting in a slower on-rate and faster off-rate.
A frequent problem encountered with nerve terminal dyes is background fluorescence due to non-vesicle membrane staining. While most background fluorescence can be removed with repeated washes, background can be a problem with more lipophilic dyes, particularly in tissue preparations. Biotium offers three agents to reduce background fluorescence of nerve terminal dyes. ADVASEP-7 is a sulfonated beta-cyclodextrin that facilites removal of dye from the cell surface during washing. Biotium's unique quencher SCAS reduces background fluorescence without additional washes. Sulforhodamine 101 has been used to quench SynaptoGreen background by fluorescence resonance energy transfer (FRET). These background reducing agents are available separately or together with commonly used styryl dyes in nerve terminal staining kits.
These dyes have a lipophilic tail at one end and a hydrophilic cationically charged head group at the other end. The dyes vary in the number of carbons in the lipophilic tail (designated m) and the number of double bonds linking the two aromatic rings in the dye (designated n). AM dyes are similar to FM dyes, but contain an additional formaldehyde-fixable amino group in the cationic head.
Ordering information: Sulforhodamine 101 is also available separately (catalog number 89139-504).