Human Recombinant IFN gamma
Supplier: IBI Scientific
RPH-101 RPH-102 RPH-103 RPH-104 RPH-105 RPH-106
71005-090EA 631.86 CAD
71005-090 71005-092 71005-094 71005-096 71005-098 71005-100
Human Recombinant IFN gamma
Proteins and Peptides
Recombinant proteins are used extensively in today's Life Science Research Laboratory. IBI proteins focus primarily on Animal Models and Veterinary Research, but also contain some Human Recombinant proteins.
IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.
IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.