Lipophilic Carbocyanine Dyes and CellBrite Cytoplasmic Membrane Labeling Kits, Biotium
Supplier: Biotium
30021 30022 30023 30024 60010 60011 60012 60013 60015 60016 60018 60019 60020 60034 60035 60036 60038
89138-942EA 352.59 CAD
89138-942 89138-944 89138-946 89138-948 89139-298 89139-300 89139-302 89139-304 89139-308 89139-310 89139-314 89139-316 89139-318 89139-342 89139-344 89139-346 89139-350
Lipophilic Carbocyanine Dyes and CellBrite Cytoplasmic Membrane Labeling Kits, Biotium
Protein Linking and Labeling Systems
Carbocyanine dyes with long hydrocarbon chains have low fluorescence in aqueous phase, but become highly fluorescent upon binding to lipid bilayers, making this class of dyes ideal for staining cell membranes
Biotium offers a series of novel and classic carbocyanine dyes with fluorescent colors ranging from blue to infrared, including the novel blue carbocyanine dye DiB, and Neuro-DiO, a green fluorescent dye with improved solubility and photostability compared to DiO. Biotium also offers ready-to-use CellBrite™ Cell Membrane Labeling Kits with fluorescent colors ranging from blue to infrared. The labeling kits offer fast and intensely bright labeling of cells without dye precipi­tation, and include a detailed labeling protocol.
Carbocyanine dye mem­brane staining is very stable, with minimal dye transfer between cells. Dilinoleyl carbocynaine dyes have much faster lateral diffusion rates than regular carbocyanine dyes, which facilitates labeling of neurons in tissues.