Connecting Adapter Kit, Multipack, Rotary Evaporator, Ace Glass Incorporated
Supplier: Ace Glass
13290-55 13290-59 13290-69
89077-632EA 561.55 CAD
89077-632 89084-018 89235-724 89235-726
Connecting Adapter Kit, Multipack, Rotary Evaporator, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This multipack contains seven chemically inert PTFE adapters with Ace-Thred and GPI vial inner thread, plus one glass conversion adapter.
  • Chemically inert
  • Seven adapters with #15 Ace-Thred and GPI vial inner thread
  • One glass conversion adapter with #15 Ace-Thred bottom and reinforced [ST] outer top joint
The seven chemically inert PTFE adapters have a #15 Ace-Thred and GPI vial inner thread, and the one glass conversion adapter has a #15 Ace-Thred bottom (that fits all of the included adapters) and a reinforced standard taper outer joint at top. These adapters are used to connect matching vials for use in rotary evaporators and are also suitable for vacuum work. The "For Use With" values represent the included glass conversion adapter.
Ordering information: Supplied with either FETFE or Chemraz o-rings, one for each of the seven PTFE adapters.
Caution: FETFE not suitable for use with methylene chloride or acetone. Use Chemraz instead.