Bottle, Threaded Neck, Ace Glass
Supplier: Ace Glass
4048-13 4048-19 4048-45 4048-09
89083-008EA 1399.27 CAD
89083-008 89083-010 89083-012 89083-014
Bottle, Threaded Neck, Ace Glass
Bottles Glass Bottles
This is a borosilicate glass bottle designed for the storage of solutions.
Bottles have a GL120 external thread finish neck with large 106 mm (4.2") Int.Ø opening. The 9.5 and 13.25 liter sizes have a conventional bottle shape. The 19 and 45.5 liter sizes are similar in design to a carboy.
Ordering information: A glass-filled PTFE cap with CAPFE (PTFE encapsulated silicone) O-ring may be purchased separately. Ask for Order Code 89059-794.
Order Now
SPECIFICATIONS
Environmentally Preferable