Beakers, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
5340-03 5340-05 5340-10 5340-15 5340-18 5340-20 5340-25 5340-30 5340-35
89051-590EA 380.34 CAD
89051-590 89051-592 89051-594 89051-596 89051-598 89051-600 89051-602 89051-604 89051-606
Beakers, Jacketed, Ace Glass Incorporated
Beakers
This jacketed beaker has one upper and one lower hose connection on opposite sides
Does not have a pouring spout. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable