Bottle, Woulff, Ace Glass Incorporated
Supplier: Ace Glass
5365-10
89051-616EA 772.81 CAD
89051-616
Bottle, Woulff, Ace Glass Incorporated
Bottles Woulff Bottles
This Woulff bottle is made of heavy wall glass, with three reinforced standard taper 24/40 outer neck joints.
Capacity is 1000 ml Fabricated from borosilicate glass.
Ordering information: Stoppers not included. For smaller capacities, contact your Avantor representative.