Beakers, PTFE, Thermotech™, Ace Glass Incorporated
Supplier: Ace Glass
5500-22 5500-24 5500-26
89082-910EA 475.37 CAD
89082-910 89082-912 89082-914
Beakers, PTFE, Thermotech™, Ace Glass Incorporated
Beakers
This pure, inert PTFE beaker has a pour spout
The bottom surface has a molded, stabilized PTFE/Carbon outer surface for better heat transfer, and to handle higher temperatures up to 270°C.