Solution Bottle, Plastic Coated, Ace Glass
Supplier: Ace Glass
5393-06 5393-09
89051-618EA 1164.37 CAD
89051-618 89051-620 89051-622
Solution Bottle, Plastic Coated, Ace Glass
Bottles Glass Bottles
This solution bottle is coated to the neck with PVC safety coating to reduce potential breakage and exposure to laboratory personnel when handling hazardous material.
  • Fabricated from borosilicate glass
This bottle is carboy style, with sloping shoulders. Necks are tooled for uniform fit with #12 rubber stoppers.
Ordering information: Stoppers are not included.