Bottle, Plastic Coated, Ace-Safe, Ace Glass
Supplier: Ace Glass
5395-103 5395-105 5395-107
89051-630EA 1515.34 CAD
89051-630 89051-632 89051-634
Bottle, Plastic Coated, Ace-Safe, Ace Glass
Bottles Glass Bottles
This solution bottle has a serrated vacuum takeoff.
  • Fabricated from borosilicate glass
  • Maximum temperature limit 80 °C
  • Bottle is carboy style, with sloping shoulders
  • Neck is tooled for uniform fit with stoppers
Takeoff has a #15 Ace-Thred for use with an Ace-Safe tubing connection barb for ³/₈" Int.Ø tubing. Bottle is coated to the neck with a plastic safety coating. This translucent coating forms a protective film around the glass that will help contain both particles and dangerous chemicals should the glass break.
Ordering information: Supplied complete with a Size F hose connection with o-ring and nylon bushing (89052-956). Stopper is NOT included. Higher temperature coatings are available via special order. Contact your Avantor representative for more information.