Beakers, Big Jar, Ace Glass Incorporated
Supplier: Ace Glass
6233-17 6233-26 6233-32 6233-07 6233-09
89082-888EA 2224.28 CAD
89082-888 89082-890 89082-892 89082-894 89082-896
Beakers, Big Jar, Ace Glass Incorporated
Beakers
This cylindrical beaker/jar has side indents for easier handling.
  • Fabricated from borosilicate glass
  • Made of heavy wall glass
  • Graduated, with pour spout
Caution: Do not apply open flame or heat.
Ordering information: Also available with poly safety coating. Contact your Avantor representative for more information and pricing.