Adapter, Stainless Steel, Circulator Hose, NPT, Ace Glass Incorporated
Supplier: Ace Glass
12187-100 12187-101 12187-102
89097-272EA 454.08 CAD
89097-272 89097-274 89097-276
Adapter, Stainless Steel, Circulator Hose, NPT, Ace Glass Incorporated
Fittings and Connectors
These stainless steel adapters are designed for use with ACE jacketed glass pilot plant reactors and circulator hoses for popular circulating chillers.
  • Bottom M style male
  • Top NPT thread
  • Stainless steel
This hose-to-circulator adapter connects “M” style to male NPT thread.