Adapter, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Supplier: Ace Glass
12187-10 12187-12 12187-14 12187-05 12187-07
71001-580EA 329.11 CAD
71001-580 71001-582 71001-584 89097-264 89097-266
Adapter, Stainless Steel, Circulator Hose, Ace Glass Incorporated
Fittings and Connectors
These stainless steel adapters are designed for use with ACE jacketed glass pilot plant reactors and circulator hoses for popular circulating chillers.
  • Bottom spherical joint
  • Top M style male
  • Stainless steel
This flask-to-hose adapter connects the o-ring ball joints on the jacket inlet/outlet to hoses to/from the circulator.