PIG® Grippy® Absorbent Mat Roll, New Pig
Supplier: New Pig
MAT1625 MAT32100 MAT3250
89212-350EA 112.65 CAD
89212-350 89212-352 89212-354
PIG® Grippy® Absorbent Mat Roll, New Pig
Spill Control Materials
Specially formulated, full-coverage adhesive backing holds tight but leaves no residue; eliminates the need for tapes and trays
No curling or bunching; lies flat to reduce trip hazards around workstations, walkways and machinery. Poly backing creates a liquidproof barrier that keeps floors completely dry. Tough, spunbond top layer stands up to foot and light-wheeled traffic as well as heavy equipment driveovers. Dark color hides grime so mat stays down longer.
When used as part of your floor safety program, high-traction surfaces such as PIG Grippy Mat have been proven to reduce slip-and-fall claims by as much as 90%. Tested and certified by the National Floor Safety Institute (NFSI).
Peels up easily for quick changeouts; reduces cleanup time for a more productive facility. Eight layers of fine-fiber polypropylene safely absorb leaks, drips and overspray of oils, coolants, solvents and water to keep liquids from being tracked around