VWR® Hazmat Spill Response Kit
Supplier: VWR
VWR300
89168-998EA 186.35 CAD
89168-998
VWR® Hazmat Spill Response Kit
Spill Control Materials
Hazmat spill response kit includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie.
  • Includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles, and one water-resistant yellow bag with tie
  • Supports compliance with 40 CFR 263.30(a) safety standard