Beakers, Griffin Low Form, Ace Glass Incorporated
Supplier: Ace Glass
12420-08 12420-10 12420-12 12420-14 12420-18 12420-20 12420-22
89085-370CS 171.96 CAD
89085-370 89085-372 89085-374 89085-376 89085-378 89085-380 89085-382 89085-384 89085-386
Beakers, Griffin Low Form, Ace Glass Incorporated
Beakers
These Griffin-style polypropylene beakers are for general laboratory use
  • Autoclavable
  • Approximate raised volume scales
  • Tapered walls for safe handling and convenient stacking
Combines a “no-drip” pouring spout with unbreakability and maximum translucency.
Ordering information: Lids are not included.