Vial Connecting Adapters, PTFE, Ace Glass Incorporated
Supplier: Ace Glass
13290-123 13290-125 13290-128 13290-13 13290-130 13290-132 13290-15 13290-18 13290-20 13290-22 13290-134 13290-121 13290-136 13290-24 13290-11 13290-26
89077-610EA 220.81 CAD
89077-610 89077-612 89077-614 89077-616 89077-618 89077-620 89077-622 89077-624 89077-626 89077-628 89082-652 89082-654 89082-656 89082-836 89082-838 89082-840
Vial Connecting Adapters, PTFE, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
Chemically inert PTFE adapters feature either a #15 or #25 Ace-Thred.
  • GPI vial inner thread
  • Suitable for vacuum work
  • O-ring included
A GPI vial inner thread connects conversion adapters to the matching vial for use in rotary evaporators. Suitable for vacuum work. Supplied with either FETFE or Chemraz O-ring. Thread Size represents the inner GPI Thread.
Caution: FETFE is not suitable for use with methylene chloride or acetone. Chemraz is suitable for use instead.