GlobalMark® Tape, B-582, Brady Worldwide®
Supplier: Brady Worldwide
76711 76712 76713
CA89007-714EA 914.71 CAD
CA89007-714 CA89007-716 CA89007-718
GlobalMark® Tape, B-582, Brady Worldwide®
Tapes
GlobalMark tapes are easy to install and very durable
Brady B-582 BradyGlo material is a glow-in-the-dark (photoluminescent) pressure sensitive polyester designed for use in the Labelizer Plus Industrial Labeling System, the GlobalMark Industrial Label Maker and the PowerMark Sign and Label Maker. In accordance with ASTM 2030-99: Recommended Uses of Photoluminescent Safety Markings. Not recommended for outdoor application. Meets or exceeds the following standards. ASTM E-2072-00 / ASTM E-2073-00, ASTM E-162, ASTM E-648, ASTM E-662, IMO Resolution A.752(18), ISO/CD 15370, Marine Safety Committee MSC.27(61), DIN 67 510 (Parts 1 - 4) and PSPA Standard 002 Part 2. Meets OSHA 1910.37. Lloyd's Register SAS F050294.p Charge time of 5 minutes at 1000 Lux will charge the sign for plus 10 hours of glow time. Consistent glow life and unlimited rechargability throughout service life. Minimum Application Temperature: 50° F (10°C).