GlobalMark® Reflective Tape, B-584, Brady Worldwide®
Supplier: Brady Worldwide
76705 76706 76707
CA89007-708EA 831.22 CAD
CA89007-708 CA89007-710 CA89007-712
GlobalMark® Reflective Tape, B-584, Brady Worldwide®
Tapes
GlobalMark tapes are easy to install and very durable
Not recommended for plotter cutting. Brady B-584 is a reflective tape designed for use with the Labelizer Plus Industrial Labeling System, GraphicsPro Label and Sign Creator, VersaPrinter Label and Sign Maker System, and the HandiMark Portable Label Maker, PowerMark Sign and Label Maker and GlobalMark Industrial Label Maker. Minimum Application Temperature: Silver: 50°F (10°C); Yellow and Orange: 0°F (-18°C).
Order Now
SPECIFICATIONS
Environmentally Preferable