Adapter. Distilling, Ace-Thred, Ace Glass Incorporated
Supplier: Ace Glass
5086-54
89051-308EA 280.58 CAD
89051-308
Adapter. Distilling, Ace-Thred, Ace Glass Incorporated
Distillation Components Distillation Adapters
This adapter is designed for connecting distilling columns with a vertical condenser.
  • Top Ace-Thred and thermometer
  • Bottom [ST] inner joints
  • Side arm at 75° angle
  • Nylon bushing and FETFE o-ring included
  • Fabricated from borosilicate glass
The top of this adapter has a #7 Ace-Thred and an adjustable length thermometer. The side arm is at 75 degrees, and the vertical side arm is 17.8cm from the center tube. Bottom joints are standard taper 24/40 inner. Supplied complete with 89051-122 nylon bushing and FETFE o-ring. For extra o-rings order 89060-958.