Adapter, Outer Socket Joint, Ace Glass Incorporated
Supplier: Ace Glass
5221-09 5221-11 5221-05 5221-24 5221-28 5221-20
89082-550EA 120.59 CAD
89082-550 89082-552 89082-554 89082-574 89082-576 89082-578
Adapter, Outer Socket Joint, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This series of adapters is designed for joining jacketed reactor ball joint inlet/outlets to recirculator hoses.
  • Outer socket joints
  • Various Ace-Thred and tube ends
  • Fabricated from borosilicate glass
Adapters feature 28/15 and 35/25 sockets with various Ace-Thred and tube ends.